N-(4-Carboxyphenyl)succinamic acid 99%
Supplier: Thermo Scientific
|
Warning
|
L17549.06
L17549.03
L17549.06EA
58
EUR
L17549.06
L17549.03
N-(4-Carboxyphenyl)succinamic acid 99%
N-(4-Carboxyphenyl)succinamic acid
Purity: 99%
CAS No.: 76475-62-2
Molecular Formula: C11H11NO5
Formula Weight: 237.21
Melting Point: ca 290° dec.
Beilstein Reference: 3355574
MDL No.: MFCD00029834
Formula:
C₁₁H₁₁NO₅ MW: 237,21 g/mol Melting Pt: ∼ 290 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00029834 CAS Number: 76475-62-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...