5-Bromo-1H-indol-7-amine 97%
Supplier: Thermo Scientific
|
Danger
|
L17795.MC
L17795.ME
L17795.03
L17795.MCEA
65.7
EUR
L17795.MC
L17795.ME
L17795.03
5-Bromo-1H-indol-7-amine 97%
5-Bromo-1H-indol-7-amine
Purity: 97%
CAS No.: 374537-99-2
Molecular Formula: C8H7BrN2
Formula Weight: 211.06
Storage Temperature: -30°C to -10°C
Melting Point: 104-105°
UN No.: UN2811
MDL No.: MFCD02093961
Formula:
C₈H₇BrN₂ Melting Pt: 104…105 °C |
MDL Number:
MFCD02093961 CAS Number: 374537-99-2 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...