4-(Methylsulphinylphenyl)boronic acid 98%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
(4-(Methylsulphinyl)phenyl)boronic acid
L17865.03
L17865.MC
L17865.03EA
246
EUR
L17865.03
L17865.MC
4-(Methylsulphinylphenyl)boronic acid 98%
4-(Methylsulphinylphenyl)boronic acid
Purity: 98%
CAS No.: 166386-48-7
Molecular Formula: C7H9BO3S
Formula Weight: 184.02
Melting Point: ca 170°
MDL No.: MFCD02093071
Formula:
C₇H₉BO₃S MW: 184,02 g/mol Melting Pt: ∼ 170 °C Storage Temperature: Ambient |
MDL Number:
MFCD02093071 CAS Number: 166386-48-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...