3-Methylthiophene-2-methylamine 96%
Supplier: Thermo Fisher Scientific
|
Danger
|
L18094.03
L18094.MD
L18094.03EA
281
EUR
L18094.03
L18094.MD
3-Methylthiophene-2-methylamine 96%
3-Methylthiophene-2-methylamine
Purity: 98%
CAS No.: 104163-35-1
Molecular Formula: C6H9NS
Formula Weight: 127.21
Refractive Index: 1.5565
Boiling Point: 78°/4mm
Beilstein Reference: 108864
UN No.: UN2735
MDL No.: MFCD02093988
Formula:
C₆H₉NS Boiling Pt: 78 °C (4 mmHg) Storage Temperature: Ambient |
MDL Number:
MFCD02093988 CAS Number: 104163-35-1 UN: 2735 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...