1-Methylpyrrole-2-carbonyl chloride 95%
Supplier: Thermo Scientific
|
Danger
|
L18431.06
L18431.03
L18431.06EA
509
EUR
L18431.06
L18431.03
1-Methylpyrrole-2-carbonyl chloride 95%
1-Methylpyrrole-2-carbonyl chloride
Purity: 95%
CAS No.: 26214-68-6
Molecular Formula: C6H6ClNO
Formula Weight: 143.57
Storage Temperature: -30°C to -10°C
Refractive Index: 1.5700
Boiling Point: 72-74°/10mm
Beilstein Reference: 472022
UN No.: UN3265
MDL No.: MFCD02094021
Formula:
C₆H₆ClNO MW: 143,57 g/mol Boiling Pt: 72…74 °C (10 mmHg) |
MDL Number:
MFCD02094021 CAS Number: 26214-68-6 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...