4-Acetamidobenzoyl chloride 95%

Supplier: Thermo Scientific
Danger

L18432.03 L18432.06
L18432.03EA 50.9 EUR
L18432.03 L18432.06
4-Acetamidobenzoyl chloride 95%
4-Acetamidobenzoyl chloride

Purity: 95% CAS No.: 16331-48-9 Molecular Formula: C9H8ClNO2 Formula Weight: 197.62 Melting Point: ca 145° dec. UN No.: UN3261 MDL No.: MFCD02094022

Formula: C₉H₈ClNO₂
MW: 197,62 g/mol
Melting Pt: ∼ 145 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD02094022
CAS Number: 16331-48-9
EINECS: 000-000-0
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR