4-((tert-Butoxycarbonyl)amino)benzoic acid 97%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
Boc-4-Abz-OH, 4-(Boc-amino)benzoic Acid
L18531.06
L18531.03
L18531.06EA
69
EUR
L18531.06
L18531.03
4-((tert-Butoxycarbonyl)amino)benzoic acid 97%
4-((tert-Butoxycarbonyl)amino)benzoic acid
Purity: 97%
CAS No.: 66493-39-8
Molecular Formula: C12H15NO4
Formula Weight: 237.26
Melting Point: ca 193° dec.
Beilstein Reference: 2115614
MDL No.: MFCD00037428
Formula:
C₁₂H₁₅NO₄ MW: 237,3 g/mol Melting Pt: ∼ 193 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00037428 CAS Number: 66493-39-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...