Boc-Thz-OH 98%
Supplier: Thermo Fisher Scientific
Synonyms:
Boc-L-thiazolidine-4-carboxylic acid
L19488.03
L19488.06
L19488.03EA
49.4
EUR
L19488.03
L19488.06
Boc-Thz-OH 98%
Boc-Thz-OH
Purity: 98%
CAS No.: 51077-16-8
Molecular Formula: C9H15NO4S
Formula Weight: 233.28
Melting Point: ca 133° dec.
Beilstein Reference: 6686084
Optical Rotation: -113° (c=1 in methanol)
MDL No.: MFCD00077002
Formula:
C₉H₁₅NO₄S Melting Pt: ∼ 133 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00077002 CAS Number: 51077-16-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...