trans-β-Styrylboronic acid pinacol ester ≥98%

Supplier: Thermo Fisher Scientific

Synonyms: E-Phenylethenylboronic acid, pinacol ester, Trans-2-Phenylvinylboronic acid pinacol ester, trans-2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)styrene

L19529.03 L19529.MD
L19529.03EA 120 EUR
L19529.03 L19529.MD
trans-β-Styrylboronic acid pinacol ester ≥98%
trans-beta-Styrylboronic acid pinacol ester

Purity: 99% CAS No.: 83947-56-2 Molecular Formula: C14H19BO2 Formula Weight: 230.12 Flash Point: 110°(230°F) Refractive Index: 1.5290 Melting Point: 27-33° Boiling Point: 128°/4mm Beilstein Reference: 8617179 MDL No.: MFCD03453666

Formula: C₁₄H₁₉BO₂
Boiling Pt: 128 °C (4 mmHg)
Melting Pt: 27…33 °C
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD03453666
CAS Number: 83947-56-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR