Amberlite XAD-7 ion exchange resin

Supplier: Thermo Scientific

L19564.22 L19564.36
L19564.22EA 46.6 EUR
L19564.22 L19564.36
Amberlite XAD-7 ion exchange resin
Amberlite XAD-7

CAS No.: 37380-43-1 MDL No.: MFCD00132705

Storage Temperature: Ambient
MDL Number: MFCD00132705
CAS Number: 37380-43-1
Merck Index: 14,382

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR