(1S)-(+)-Menthyl glyoxylate monohydrate ≥98%
Supplier: Thermo Scientific
|
Danger
|
L19785.14
L19785.22
L19785.14EA
78.7
EUR
L19785.14
L19785.22
(1S)-(+)-Menthyl glyoxylate monohydrate ≥98%
(1S)-(+)-Menthyl glyoxylate monohydrate
Purity: 98%
Molecular Formula: C12H22O4
Formula Weight: 230.31 (212.29anhy)
Melting Point: 76-80°
Boiling Point: 95-96°/0.3mm
Beilstein Reference: 4905777
Optical Rotation: +74° (c=1 in methanol)
UN No.: UN3077
MDL No.: MFCD04037548
Formula:
C₁₂H₂₂O₄ MW: 230,31 g/mol (212,29 anhy) Boiling Pt: 95…96 °C (0,3 mmHg) Melting Pt: 76…80 °C Density: 1,182 Storage Temperature: Ambient |
MDL Number:
MFCD04037548 ADR: 9,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...