1-Trityl-(R)-3-methylpiperazine 98%, ee 99%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
(R)-3-Methyl-1-tritylpiperazine
L20078.03
L20078.MD
L20078.03EA
200
EUR
L20078.03
L20078.MD
1-Trityl-(R)-3-methylpiperazine 98%, ee 99%
1-Trityl-(R)-3-methylpiperazine
Purity: 98%
CAS No.: 313657-75-9
Molecular Formula: C24H26N2
Formula Weight: 342.49
Melting Point: 128-129°
Optical Rotation: -7.6° (c=1 in MeOH)
MDL No.: MFCD03426411
Formula:
C₂₄H₂₆N₂ MW: 342,49 g/mol Melting Pt: 128…129 °C Storage Temperature: Ambient |
MDL Number:
MFCD03426411 CAS Number: 313657-75-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...