3-Methoxyphenylglyoxal hydrate 97% (dry weight)

Supplier: Thermo Scientific
Warning

Synonyms: (3-Methoxyphenyl)(oxo)acetaldehyde

L20263.03 L20263.06
L20263.03EA 76.2 EUR
L20263.03 L20263.06
3-Methoxyphenylglyoxal hydrate 97% (dry weight)
3-Methoxyphenylglyoxal hydrate

Purity: 97% CAS No.: 32025-65-3 Molecular Formula: C9H8O3.xH2O Formula Weight: 164.16(anhy) Melting Point: 94-98° Boiling Point: 95-98°/0.05mm Beilstein Reference: 1935225 MDL No.: MFCD05664093

Formula: C₉H₈O₃·xH₂O
MW: 164,16 g/mol (anhy)
Boiling Pt: 95…98 °C (0,05 mmHg)
Melting Pt: 94…98 °C
Storage Temperature: Ambient
MDL Number: MFCD05664093
CAS Number: 32025-65-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR