Pyridine-4-boronic acid hydrate, (H₂O 10-15%)

Supplier: Thermo Fisher Scientific
Warning

L15179.03 L15179.06
L15179.03EA 0 EUR
L15179.03 L15179.06
Pyridine-4-boronic acid hydrate, (H₂O 10-15%)
Pyridine-4-boronic acid hydrate

CAS No.: 1692-15-5 Molecular Formula: C5H6BNO2.xH2O Formula Weight: 122.92(anhy) Storage Temperature: 2°C to 8°C Melting Point: 300° Beilstein Reference: 471944

Formula: C₅H₆BNO₂·xH₂O
Melting Pt: >300 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08061353
CAS Number: 1256355-27-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR