4-Chloro-3,5-xylenol USP
Supplier: Spectrum Chemical
|
Warning
|
Synonyms:
2-Chloro-5-hydroxy-3-xylene, Chloroxylenol, 4-Chloro-3,5-dimethylphenol
CH136-12KGBL
CH136-25KGBL
CH136-50KGBL
SPCMCH136-12KGBLEA
0
EUR
SPCMCH136-12KGBL
SPCMCH136-25KGBL
SPCMCH136-50KGBL
4-Chloro-3,5-xylenol USP
4-Chloro-3,5-xylenol
BSE/TSE statement (available upon request)
Allergen statement (available upon request)
Formula:
C₈H₉ClO MW: 156.61 g/mol Boiling Pt: 246 Melting Pt: 114…116 °C Density: 1.364 g/cm³ (20 °C) Flash Pt: 132 |
MDL Number:
MFCD00002324 CAS Number: 88-04-0 EINECS: 201-793-8 Merck Index: 13,02194 |
Specification Test Results
Assay | 98.5% Min. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...