Avidin, horseradish peroxidase (HRP) conjugated, Pierce™
Supplier: Thermo Fisher Scientific
29994 21123
PIER29994EA 424 EUR
PIER29994 PIER21123
Avidin, horseradish peroxidase (HRP) conjugated, Pierce™
Protein Linking and Labeling Systems
Pierce™ HRP conjugated avidin is used in biotin-based detection methods, including Western blotting or ELISA detection with appropriate enzyme substrates.
  • Native avidin - glycoprotein from chicken egg whites, 67 kDa, pI = 10
  • Glycosylation and isoelectric point renders avidin more soluble than streptavidin
  • Significantly less expensive than recombinant streptavidin
  • Avidin is usually preferred over streptavidin for avidin-biotin complex (ABC) staining methods in immunohistochemistry (IHC)
  • HRP conjugate ideal for membrane and plate-based assays
Order Now
SPECIFICATIONS
Environmentally Preferable
Host
Learn more