Anti-IgG Llama Polyclonal Antibody (CF™ 488A)

Supplier: Biotium
20454-50UL 20454-500UL
20454-50ULEA 127 EUR
20454-50UL 20454-500UL
Anti-IgG Llama Polyclonal Antibody (CF™ 488A)
Antibodies
Llama anti-mouse CF™ dye conjugates are prepared by labeling affinity purified llama anti-mouse IgG (H+L) with a selection of CF™ dyes. CF™ dyes offer combined advantages in brightness, photostability, and specificity compared other fluorescent dyes. CF™488A (Ex/Em 490/515 nm) is a superior alternative to the traditional green fluorescent dye FITC (or fluorescein) owing to its exceptional brightness and photostability. CF™488A has brightness and photostability comparable to Alexa Fluor® 488.

Alexa Fluor® is a registered trademark of Thermo Fisher Scientific.

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: CF488A
Epitope:
Host: Llama
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR