Bis(phenylsulphonyl)sulphide 96%

Supplier: Thermo Scientific

Synonyms: Benzenesulphonic thioanhydride

300830010 300830050
ACRO300830010EA 110 EUR
ACRO300830010 ACRO300830050
Bis(phenylsulphonyl)sulphide 96%
Bis(phenylsulphonyl)sulphide

CAS No.: 4388-22-1

Formula: C₁₂H₁₀O₄S₃
MW: 314,388 g/mol
Melting Pt: 129…134 °C
Storage Temperature: Ambient
MDL Number: MFCD00094733
CAS Number: 4388-22-1

Order Now

Specification Test Results

Appearance Off-white to pink Powder or crystals
Infrared spectrum Conforms
Melting point 129 to 134 °C
HPLC ≥95 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR