Amberlite® HPR900 (OH) resin_ion-exchange

Supplier: Thermo Fisher Scientific
Danger

301341000 301345000 301340025
ACRO301341000EA 33.5 EUR
ACRO301341000 ACRO301345000 ACRO301340025
Amberlite® HPR900 (OH) resin_ion-exchange
Amberlite® HPR900 (OH)

CAS No.: 197809-12-4

Storage Temperature: Ambient
MDL Number: MFCD00802695
CAS Number: 197809-12-4

Order Now

Specification Test Results

Appearance Cream Beads
Particle size ≤5.0 % (> 1.180 mm)
Harmonic mean size 0.560 to 0.700 mm
Uniformity coefficient ≤1.45
Moisture holding capacity 66.0 to 75.0 % (OH)
Total volume capacity ≥0.9 eq/l (OH)
Impurity ≤1.0 % Chloride sites content


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR