4-Aminostyrene 90% stabilised, Technical Grade

Supplier: Thermo Fisher Scientific
Danger

Synonyms: 4-Vinylaniline

308480050 308480010 308480250
ACRO308480050EA 177 EUR
ACRO308480050 ACRO308480010 ACRO308480250
4-Aminostyrene 90% stabilised, Technical Grade
4-Aminostyrene

Appearance: Yellow to orange-red Liquid or solid

Formula: C₈H₉N
MW: 119.17 g/mol
Boiling Pt: 249 °C (760 mmHg)
Melting Pt: 23…24 °C
Density: 1.01 g/cm³
Flash Pt: 99 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00015329
CAS Number: 1520-21-4
UN: 2811
ADR: 6.1 III

Order Now

Specification Test Results

Appearance Yellow to orange-red Liquid or solid
Infrared spectrum Conforms
GC ≥88 %
Stabilizer Approx. 100 ppm 4-tert-Butylcatechol


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR