4-Fluorobenzyl isocyanate 98%
Supplier: Thermo Fisher Scientific
|
Warning
|
319300010
319300050
ACRO319300010EA
92.9
EUR
ACRO319300010
ACRO319300050
4-Fluorobenzyl isocyanate 98%
4-Fluorobenzyl isocyanate
Appearance: Colorless Liquid
Formula:
C₈H₆FNO Boiling Pt: 211 °C (760 mmHg) Density: 1,186 g/cm³ Flash Pt: 93 °C Storage Temperature: Freezer |
MDL Number:
MFCD00673062 CAS Number: 132740-43-3 UN: 2206 ADR: 6.1,III |
Specification Test Results
Appearance | Colorless Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5040 to 1.5060 (20°C, 589 nm) |
GC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...