Thapsigargin 97%
Supplier: Thermo Fisher Scientific
|
Danger
|
328570010
328570050
ACRO328570010EA
245
EUR
ACRO328570010
ACRO328570050
Thapsigargin 97%
Thapsigargin
CAS No.: 67526-95-8
Formula:
C₃₄H₅₀O₁₂ Storage Temperature: Freezer |
MDL Number:
MFCD00083511 CAS Number: 67526-95-8 |
Specification Test Results
Appearance | Colorless Film |
HPLC | ≥96.0 % |
TLC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...