2-Methyl-DL-serine hydrate 96%
Supplier: Thermo Scientific
Synonyms:
2-Methylserine hydrate, (R,S)-2-Amino-3-hydroxy-2-methylpropionic acid hydrate
339750010
339750050
ACRO339750010EA
107
EUR
ACRO339750010
ACRO339750050
2-Methyl-DL-serine hydrate 96%
2-Methyl-DL-serine hydrate
CAS No.: 5424-29-3
Formula:
C₄H₉NO₃.xH₂O MW: 119,12 g/mol (anhy) Melting Pt: 253…263 °C Storage Temperature: Ambient |
MDL Number:
MFCD03791288 CAS Number: 5424-29-3 EINECS: 226-565-5 |
Specification Test Results
Appearance | White to off-white Powder |
Infrared spectrum | Conforms |
Water | ≤10 % (K.F.) |
Titration with HClO4 | ≥95 % (On dry substance) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...