3-Amino-2,2,5,5-tetramethyl-1-pyrrolidinyloxy 99%

Supplier: Thermo Fisher Scientific
Danger

401105000 401100020
ACRO401105000EA 210 EUR
ACRO401105000 ACRO401100020
3-Amino-2,2,5,5-tetramethyl-1-pyrrolidinyloxy 99%
3-Amino-2,2,5,5-tetramethyl-1-pyrrolidinyloxy

Appearance: Orange Hygroscopic crystals

Formula: C₈H₁₇N₂O
Boiling Pt: 75…80 °C (1 mmHg)
Melting Pt: 45 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00046088
CAS Number: 34272-83-8
UN: 3263
ADR: 8,III

Order Now

Specification Test Results

Appearance Orange Hygroscopic crystals
Infrared spectrum Conforms
Titration with HClO4 ≥98.5 %
GC ≥97.5 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR