3-Bromobutanone 95% stabilised

Supplier: Thermo Scientific
Danger

402910050 402910010
ACRO402910050EA 117 EUR
ACRO402910050 ACRO402910010
3-Bromobutanone 95% stabilised
3-Bromobutanone
Formula: H₃CC(O)BrCHCH₃
MW: 151 g/mol
Boiling Pt: 36 °C (11 mmHg)
Density: 1,438 g/cm³
Flash Pt: 51 °C
Storage Temperature: Ambient
MDL Number: MFCD00013538
CAS Number: 814-75-5
UN: 2920
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear light yellow to brown Liquid
Infrared spectrum Conforms
Refractive index 1.4580 to 1.4600 (20°C, 589 nm)
GC ≥94 %
Stabilizer ≤1 % magnesium oxide

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR