p-Hydrazinobenzenesulphonic acid hemihydrate 98%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: Phenylhydrazine-4-sulphonic acid, 4-Hydrazinobenzenesulphonic acid, hemihydrate

411780500 411781000
ACRO411780500EA 103 EUR
ACRO411780500 ACRO411781000
p-Hydrazinobenzenesulphonic acid hemihydrate 98%
p-Hydrazinobenzenesulphonic acid hemihydrate

Appearance: Light yellow to light orange Powder

Formula: C₆H₈N₂O₃S
Melting Pt: 285 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00480958
CAS Number: 854689-07-9

Order Now

Specification Test Results

Appearance Light yellow to light orange Powder
Infrared spectrum Conforms
Loss on drying 4 % to 5 %
Titration with NaOH ≥97.5 %
Titration with HClO4 ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR