Sulphorhodamine B acid chloride, pure

Supplier: Thermo Fisher Scientific
Warning

Synonyms: Lissamine rhodamine B sulphonyl chloride

413230010 413230050
ACRO413230010EA 235 EUR
ACRO413230010 ACRO413230050
Sulphorhodamine B acid chloride, pure
Sulphorhodamine B acid chloride

Appearance: Dark green Crystalline powder

Formula: C₂₇H₂₉ClN₂O₆S₂
MW: 577.107 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00042001
CAS Number: 62796-29-6

Order Now

Specification Test Results

Appearance Dark green Crystalline powder
Infrared spectrum Conforms
UV Lambda max 561 to 567 nm (in CH2Cl2)
Molar extinction coefficient ≥85000 l/(mol.cm) (lambda max)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR