Sodium tetradecyl sulphate 95%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
Tetradecyl sulphuric acid salt, Tetradecyl sulphate sodium, Sodium 1-tetradecyl sulphate
420260050
420260250
ACRO420260050EA
140
EUR
ACRO420260050
ACRO420260250
Sodium tetradecyl sulphate 95%
Sodium tetradecyl sulphate
CAS No.: 1191-50-0
Formula:
C₁₄H₂₉NaO₄S MW: 316,44 g/mol Melting Pt: 199 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007468 CAS Number: 1191-50-0 |
Specification Test Results
Appearance | White to off-white Crystalline powder |
Infrared spectrum | Conforms |
Assay | ≥94 % |
Additional info | Cr2O3: 0.00005 % |
Particle size | 205 µm (D50) Typical |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...