4-(Chloromethyl)styrene 90% tech.

Supplier: Thermo Fisher Scientific
Danger

Synonyms: 1-(Chloromethyl)-4-vinylbenzene, 4-Vinylbenzyl Chloride

422521000 422520250 422525000
ACRO422521000EA 164 EUR
ACRO422521000 ACRO422520250 ACRO422525000
4-(Chloromethyl)styrene 90% tech.
4-(Chloromethyl)styrene

Appearance: Clear yellow Liquid

Formula: C₉H₉Cl
Density: 1,08 g/cm³
Flash Pt: 105 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00051362
CAS Number: 1592-20-7
UN: 2922
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear yellow to dark yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5740 to 1.5760 (20°C, 589 nm)
GC ≥88 %
Stabilizer About 500 ppm Nitrocresol + Nitrophenol


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR