2-Chloroacrylonitrile 99% Stabilised
Supplier: Thermo Fisher Scientific
|
Danger
|
108560050
108560250
ACRO108560050EA
49.7
EUR
ACRO108560050
ACRO108560250
2-Chloroacrylonitrile 99% Stabilised
2-Chloroacrylonitrile
Appearance: Clear colorless to yellow Liquid
Formula:
C₃H₂ClN MW: 87.506 g/mol Boiling Pt: 88…89 °C (760 mmHg) Melting Pt: –65 °C Density: 1.096 g/cm³ Flash Pt: 6 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001858 CAS Number: 920-37-6 UN: 3273 ADR: 3 II |
Specification Test Results
Appearance | Clear colorless to yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4290 to 1.4310 (20°C, 589 nm) |
GC | ≥98.5 % |
Stabilizer | ca. 500 ppm Hydroquinone |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...