Pargyline 97%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: N-Methyl-N-propargylbenzylamine, N-Benzyl-N-methylprop-2-yn-1-amine

127530010 127530050
ACRO127530010EA 27.2 EUR
ACRO127530010 ACRO127530050
Pargyline 97%
Pargyline

Appearance: Clear yellow Liquid

Formula: C₁₁H₁₃N
MW: 159.232 g/mol
Boiling Pt: 86…88 °C (4 mmHg)
Density: 0.94 g/cm³
Flash Pt: 83 °C
Storage Temperature: Ambient
MDL Number: MFCD00008576
CAS Number: 555-57-7
UN: 2810
ADR: 6.1 III

Order Now

Specification Test Results

Appearance Clear yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5210 to 1.5230 (20°C, 589 nm)
Titration with HClO4 ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR