1-Acetamidoadamantane 99+%

Supplier: Thermo Fisher Scientific
Warning

133600250 133601000
ACRO133600250EA 77 EUR
ACRO133600250 ACRO133601000
1-Acetamidoadamantane 99+%
1-Acetamidoadamantane

Appearance: White to yellow Crystalline powder

Formula: C₁₂H₁₉NO
Melting Pt: 148…150 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00074730
CAS Number: 880-52-4

Order Now

Specification Test Results

Appearance White to yellow Crystalline powder
Infrared spectrum Conforms
Melting point 146 to 149 °C
GC ≥99.0 %
Elemental Analysis 52.4 to 56.8 % C
Specific optical rotation -40° to -50° (20 °C, 589 nm) (c=1 in methanol)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR