2-Piperidineethanol 95%

Supplier: Thermo Scientific
Danger

Synonyms: 2-(2-Hydroxyethyl)piperidine

157611000 157615000 157610050
ACRO157611000EA 284 EUR
ACRO157611000 ACRO157615000 ACRO157610050
2-Piperidineethanol 95%
2-Piperidineethanol

Appearance: White to light yellow Low melting crystalline solid

Formula: C₇H₁₅NO
MW: 129,20 g/mol
Boiling Pt: 234 °C (760 mmHg)
Melting Pt: 38…43 °C
Density: 1,01 g/cm³
Flash Pt: 102 °C
Storage Temperature: Ambient
MDL Number: MFCD00005989
CAS Number: 1484-84-0
UN: 3259
ADR: 8,III

Order Now

Specification Test Results

Appearance White to light yellow Low melting crystalline solid
Infrared spectrum Conforms
Melting point 38 to 43 °C
GC ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR