Pregnenolone 99%
Supplier: Thermo Fisher Scientific
164420050
164420250
164421000
ACRO164420050EA
82.2
EUR
ACRO164420050
ACRO164420250
ACRO164421000
Pregnenolone 99%
Pregnenolone
CAS No.: 145-13-1
Formula:
C₂₁H₃₂O₂ MW: 316.485 g/mol Melting Pt: 190…193 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003628 CAS Number: 145-13-1 |
Specification Test Results
Appearance | White to off-white Powder |
Infrared spectrum | Conforms |
Loss on drying | ≤0.5 % |
GC | ≥98.5 % |
Specific optical rotation | +27° to +30° (20°C, 589 nm) (c=1, EtOH) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...