Zein, purified

Supplier: Thermo Scientific

179311000 179315000 179310025
ACRO179311000EA 68.9 EUR
ACRO179311000 ACRO179315000 ACRO179310025
Zein, purified
Zein

CAS No.: 9010-66-6

Storage Temperature: Ambient
MDL Number: MFCD00062414
CAS Number: 9010-66-6

Order Now

Specification Test Results

Appearance Yellow to straw yellow to tan Powder
Infrared spectrum Conforms
Loss on drying ≤8.0 % (105°C, 2 h)
Heavy metals ≤20 ppm
Protein 88.0 to 96.0 % (On dried basis)
Residue after ignition ≤2.0 % (800°C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR