3-Phenoxypropan-1-ol 97%

Supplier: Thermo Fisher Scientific
Warning

186980050 186980250
ACRO186980050EA 55.8 EUR
ACRO186980050 ACRO186980250
3-Phenoxypropan-1-ol 97%
3-Phenoxypropan-1-ol

Appearance: Clear colorless Liquid

Formula: C₉H₁₂O₂
Boiling Pt: 83…85 °C
Density: 1,04 g/cm³
Flash Pt: >112 °C
Storage Temperature: Ambient
MDL Number: MFCD00039549
CAS Number: 6180-61-6

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.5280 to 1.5300 (20°C, 589 nm)
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR