Nigrosine, water soluble, high purity biological stain
Supplier: Thermo Fisher Scientific
189480250
189481000
ACRO189480250EA
33.4
EUR
ACRO189480250
ACRO189481000
Nigrosine, water soluble, high purity biological stain
Nigrosine
CAS No.: 8005-03-6
Storage Temperature:
Ambient |
MDL Number:
MFCD00044681 CAS Number: 8005-03-6 |
Specification Test Results
Appearance | Black Crystalline powder or crystals |
Infrared spectrum | Conforms |
UV spectrum | Complies |
HPLC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...