Potassium hexacyanocobaltate(III) 95%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: Tripotassium hexacyanocobaltate(III)

203920050 203921000 203925000
ACRO203920050EA 24.7 EUR
ACRO203920050 ACRO203921000 ACRO203925000
Potassium hexacyanocobaltate(III) 95%
Potassium hexacyanocobaltate(III)

Appearance: Light yellow to light brown Crystalline solid

Formula: C₆CoK₃N₆
Density: 1,87 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011391
CAS Number: 13963-58-1
ADR: 6.1,II

Order Now

Specification Test Results

Appearance Light yellow to light brown Crystalline solid
Assay ≥94 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR