Dicoumarol 99%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
3,3'-Methylenebis(4-hydroxycoumarin), Bishydroxycoumarin, Dicoumarin
204120050
204120250
ACRO204120050EA
39.4
EUR
ACRO204120050
ACRO204120250
Dicoumarol 99%
Dicoumarol
Appearance: White Crystalline powder
Formula:
C₁₉H₁₂O₆ MW: 336,30 g/mol Melting Pt: 290…292 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006857 CAS Number: 66-76-2 UN: 2811 ADR: 6.1,III |
Specification Test Results
Appearance | White Crystalline powder |
Infrared spectrum | Conforms |
Titration with TBAH | ≥98.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...