Diethylstilbestrol 99%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
(E)-3,4-Bis(4-hydroxyphenyl)-3-hexene, Stilbestrol, DES, (E)-4,4'-(Hex-3-ene-3,4-diyl)diphenol
204130050
204130100
204130010
ACRO204130050EA
235
EUR
ACRO204130050
ACRO204130100
ACRO204130010
Diethylstilbestrol 99%
Diethylstilbestrol
Appearance: White to off-white Crystalline powder
Formula:
C₁₈H₂₀O₂ MW: 268,356 g/mol Melting Pt: 169…175 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002373 CAS Number: 56-53-1 |
Specification Test Results
Appearance | White to off-white Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 169 to 175 °C |
HPLC | ≥98.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...