(S)-β-Phenylalaninol 95%, 98% ee

Supplier: Thermo Scientific
Danger

Synonyms: (S)-3-Amino-3-phenyl-1-propanol, (S)-3-Amino-3-phenylpropan-1-ol

362172500 362170010
ACRO362172500EA 85.9 EUR
ACRO362172500 ACRO362170010
(S)-β-Phenylalaninol 95%, 98% ee
(S)-β-Phenylalaninol
Formula: C₉H₁₃NO
MW: 151,2056 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01311768
CAS Number: 82769-76-4
UN: 3259
ADR: 8,III

Order Now

Specification Test Results

Appearance Colorless to white Viscous liquid or low melting solid
Infrared spectrum Conforms
GC ≥94 %
Enantiomeric excess ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR