Methyl-2-(bromomethyl)acrylate 96%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
2-(Bromomethyl)acrylic Acid Methyl Ester, Methyl 2-(bromomethyl)acrylate
367630010
367630050
367630250
ACRO367630010EA
220
EUR
ACRO367630010
ACRO367630050
ACRO367630250
Methyl-2-(bromomethyl)acrylate 96%
Methyl-2-(bromomethyl)acrylate
Appearance: Clear colorless to light brown Liquid
Formula:
C₅H₇BrO₂ MW: 179,01 g/mol Boiling Pt: 35…37 °C (13 mmHg) Density: 1,489 g/cm³ Flash Pt: 78 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00011697 CAS Number: 4224-69-5 UN: 3334 ADR: 9,III |
Specification Test Results
Appearance | Clear colorless to light brown Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4870 to 1.4910 (20°C, 589 nm) |
GC | ≥95 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...