3-Methyl-2-butenal 97%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
3,3-Dimethylacrolein, 3-Methylbut-2-enal, Senecialdehyde, 3-Methylcrotonaldehyde
370180050
370180250
370181000
ACRO370180050EA
64.9
EUR
ACRO370180050
ACRO370180250
ACRO370181000
3-Methyl-2-butenal 97%
3-Methyl-2-butenal
CAS No.: 107-86-8
Formula:
C₅H₈O MW: 84,12 g/mol Boiling Pt: 132…133 °C Density: 0,88 g/cm³ Flash Pt: 33 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00010291 CAS Number: 107-86-8 UN: 2920 ADR: 8,II |
Specification Test Results
Appearance | Clear colorless to yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4600 to 1.4640 (20°C, 589 nm) |
GC | ≥96.0 % |
Learn more
![](/m-ie.vwr.com/en_IE/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...