1-Methylcyclohexene 98+% stabilised
Supplier: Thermo Fisher Scientific
|
Danger
|
156840100
156840500
ACRO156840100EA
68
EUR
ACRO156840100
ACRO156840500
1-Methylcyclohexene 98+% stabilised
1-Methylcyclohexene
Formula:
C₇H₁₂ Boiling Pt: 110 °C (1013 hPa) Melting Pt: –120,4 °C Density: 0,81 g/cm³ Flash Pt: –3 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001548 CAS Number: 591-49-1 UN: 1993 ADR: 3,II |
Specification Test Results
Appearance | Clear colorless Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4492 to 1.4512 (20°C, 589 nm) |
GC | ≥98.0 % |
Stabilizer | 10 ppm hydroquinone |
Sulfate (SO4) | ≤0.015 % |
Ammonium (NH4) | ≤0.001 % |
Iron (Fe) | ≤0.001 % |
Heavy metals (as Pb) | ≤5 ppm |
Insoluble matter | ≤0.005 % (in water) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...