5,5'-Methylenedisalicylic acid 95%

Supplier: Thermo Scientific

351730050 351730250
ACRO351730050EA 44 EUR
ACRO351730050 ACRO351730250
5,5'-Methylenedisalicylic acid 95%
5,5'-Methylenedisalicylic acid

Appearance: Cream Powder

Formula: C₁₅H₁₂O₆
Melting Pt: 235…242 °C
Storage Temperature: Ambient
MDL Number: MFCD00016506
CAS Number: 122-25-8

Order Now

Specification Test Results

Appearance Cream Powder
Infrared spectrum Conforms
Melting point 232 to 241 °C
Titration with NaOH ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR