4-tert-Butylphenylboronic acid 97%
Supplier: Thermo Fisher Scientific
|
Warning
|
358970050
358970250
ACRO358970050EA
108
EUR
ACRO358970050
ACRO358970250
4-tert-Butylphenylboronic acid 97%
4-tert-Butylphenylboronic acid
Formula:
C₁₀H₁₅BO₂ Melting Pt: 191…196 °C Storage Temperature: Ambient |
MDL Number:
MFCD01009697 CAS Number: 123324-71-0 |
Specification Test Results
Appearance | White to off-white Crystalline powder |
Infrared spectrum | Conforms |
HPLC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...