N-(tert-Butoxycarbonyl)-L-propargylglycine 95%

Supplier: Thermo Scientific
Warning

Synonyms: (S)-2-((tert-Butoxycarbonyl)amino)pent-4-ynoic acid, N-Boc-L-propargylglycine, (S)-N-BOC-Propargylglycine, Boc-propargyl-Gly-OH, (S)-2-(Boc-amino)-4-pentynoic Acid, Boc-Pra-OH, (S)-2-(tert-Butoxycarbonylamino)-4-pentynoic Acid

361800010 361805000
ACRO361800010EA 315 EUR
ACRO361800010 ACRO361805000
N-(tert-Butoxycarbonyl)-L-propargylglycine 95%
N-(tert-Butoxycarbonyl)-L-propargylglycine

Appearance: Off-white to light yellow Crystalline powder

Formula: C₁₀H₁₅NO₄
MW: 213,23 g/mol
Melting Pt: 85,5 °C
Storage Temperature: Ambient
MDL Number: MFCD01320855
CAS Number: 63039-48-5

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
GC ≥94 %
Specific optical rotation +24° to +28° (20°C, 589 nm) (C=1, Methanol)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR