(R)-2-Isobutylsuccinic acid-1-methyl ester ≥95%, ee 98%

Supplier: Thermo Scientific
Warning

363050010 363055000
ACRO363050010EA 397 EUR
ACRO363050010 ACRO363055000
(R)-2-Isobutylsuccinic acid-1-methyl ester ≥95%, ee 98%
(R)-2-Isobutylsuccinic acid-1-methyl ester

Appearance: Yellow Viscous liquid

Formula: C₉H₁₆O₄
Storage Temperature: Refrigerator
MDL Number: MFCD01091032
CAS Number: 130165-76-3

Order Now

Specification Test Results

Appearance Yellow Viscous liquid
Infrared spectrum Conforms
GC ≥94 %
Enantiomeric excess ≥97.5 % (GC)
Specific optical rotation +18° to +21° (25°C, 589 nm) (c=1, CH3OH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR