Methyl-3-(dimethylamino)-2-isocyanoacrylate 96%

Supplier: Thermo Scientific
Warning

Synonyms: Methyl 3-(dimethylamino)-2-isocyanoacrylate

372020010 372020050
ACRO372020010EA 154 EUR
ACRO372020010 ACRO372020050
Methyl-3-(dimethylamino)-2-isocyanoacrylate 96%
Methyl-3-(dimethylamino)-2-isocyanoacrylate

Appearance: Yellow Crystalline powder

Formula: C₇H₁₀N₂O₂
MW: 154,169 g/mol
Melting Pt: 61…67 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04117542
CAS Number: 113212-14-9
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Yellow Crystalline powder
Infrared spectrum Conforms
Melting point 61 to 67 °C
GC ≥95 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR