Dimidium bromide 98%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
2,7-Diamino-10-methyl-9-phenylphenanthridinium bromide, 3,8-Diamino-5-methyl-6-phenylphenanthridinium bromide
215342500
215340010
ACRO215342500EA
147
EUR
ACRO215342500
ACRO215340010
Dimidium bromide 98%
Dimidium bromide
Formula:
C₂₀H₁₈BrN₃ MW: 380,29 g/mol Melting Pt: 230 °C (dec.) Storage Temperature: Refrigerator |
MDL Number:
MFCD00011757 CAS Number: 518-67-2 ADR: 6.1,I |
Specification Test Results
Appearance | Dark red to purple Powder |
Infrared spectrum | Conforms |
Titration Argentometric | ≥97.5 % (on dry substance) |
Water | ≤5 % (K.F.) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...